Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
UN:1155
ADR:3,I
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:200-467-2
REACH:01-2119535785-29
:GHS02
:GHS07
Specification Test Results
- Acidity:passes test
- Aldehydes:passes test
- Distillation Range:34.0 - 35.0 °C
- Non-volatile Matter:max. 20 mg/l
- Peroxide:passes test
- Relative Density:0.714 - 0.716
- Substances with a Foreign Odour:passes test
- Water (H2O):max. 2 g/l
Specification Test Results
Frequently Bought Together