Diisopropyl ether stabilised for synthesis, Sigma-Aldrich®
: Merck
: Isopropyl ether
:108-20-3
:
:(CH₃)₂CHOCH(CH₃)₂
:MFCD00008880
:68.5
:0.726 g/cm³ (20 °C)
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
:102.18 g/mol
:–86 °C
:–28 °C
:203-560-6
:1159
:3 II
:13,05232
:M-Clarity™ quality level = MQ200
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Solubility:12 g/l (20 °C)
- Melting Point:-86 °C
- Molar Mass:102.18 g/mol
- Boiling Point:67 - 70 °C (1013 hPa)
- Vapor Pressure:175 hPa (20 °C)
- Flash Point:-22 °C
- Refractive Index:1.3679 (20 °C, 589 nm)
- Explosion Limit:1.0 - 21 %(V)
- Density:0.72 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:405 °C
- Saturation concentration:751 g/m3 (20 °C) Air
- Evaporation number:1.6
- Water absorption:60 g/kg (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Frequently Bought Together