Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Health
Irritant
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
Dichloromethane ≥99.8% stabilised, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99.8 %
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 10 ppm
  • Water:
    Max. 0.05 %
  • Absorbance (240 nm):
    Max. 0.3
  • Absorbance (245 nm):
    Max. 0.1
  • Absorbance (260 nm):
    Max. 0.01
  • Transmittance (240 nm):
    Min. 50 %
  • Transmittance (245 nm):
    Min. 80 %
  • Transmittance (260 nm):
    Min. 98 %

Specification Test Results

Frequently Bought Together