

Dichloromethane ≥99.9% stabilised, PESTINORM® for capillary GC analysis
Catalog # 83961.320
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.9 %
- Acidity:Max. 0.0005 meq/g
- Evaporation residue (100°C):Max. 0.0005 %
- Water:Max. 0.05 %
- Organic residue (as Octanol) (GC/FID):Max. 10 ng/l
- Halogenated residue (as Lindane)(GC/ECD):Max. 5 ng/l
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
ADR:6.1,III
:Filtered through a 0.2 µm filter, packaged under nitrogen.
:Stabilised with 2-methyl-2-butene (iso-amylene) 20 ppm
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
REACH:01-2119480404-41
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881