Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate)

Tween® 20 (Polysorbate)

1
Catalog # CAYM400035-1
Supplier:   Cayman Chemical
Tween® 20 (Polysorbate)
Catalog # CAYM400035-1
Supplier:  Cayman Chemical
CAS Number:  9005-64-5
Tween® 20 (Polysorbate)

1

CAS Number:  9005-64-5
Price
23,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specifications

  • Pk:
    1

Specifications