Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate), Millipore®
Tween® 20 (Polysorbate), Millipore®
655205-250
:
Merck Millipore (Calbiochem)
:
Tween® 20 (Polysorbate), Millipore®
655205-250
:
Merck Millipore (Calbiochem)
:
Pack type:
Glass bottle
Pk:
250 mL
Seq:
2590
:
:
1227.53 g/mol
:
1.1 g/cm³ (20 °C)
:
Ambient
:
500-018-3
:
9005-64-5
:
C₅₈H₁₁₄O₂₆
:
MFCD00165986