Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 20 (Polysorbate), Proteomics Grade

Tween® 20 (Polysorbate), Proteomics Grade

Plastic bottle, 1 L
Catalog # M147-1L
Supplier:   VWR Chemicals
Tween® 20 (Polysorbate), Proteomics Grade
Catalog # M147-1L
Supplier:  VWR Chemicals
CAS Number:  9005-64-5
Tween® 20 (Polysorbate), Proteomics Grade
Tween® 20 (Polysorbate), Proteomics Grade

Plastic bottle, 1 L

CAS Number:  9005-64-5
Price
111,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Non-ionic detergent efficiently separates hydrophilic proteins from membrane spanning, hydrophobic proteins without altering biological activity.

  • High purity and protease-free
  • Suitable for most biological applications

Non-ionic detergents have uncharged hydrophilic groups and are gentle, non-denaturing detergents ideal for studying the conformation and function of proteins, because they keep secondary and tertiary structure intact.

Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1,1 g/cm³ (20 °C)
  • Flash Pt:149 °C (closed cup)
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Arsenic:
    <0,0003 %
  • DNase:
    None detected
  • Heavy metals:
    <0,001 %
  • Hydroxyl Number:
    96 - 108
  • Identification:
    Passes
  • Moisture (KF):
    <3,0 %
  • Protease:
    None detected
  • Residue on Ignition:
    ≤0,25 %
  • RNase:
    None detected

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications