Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Glass bottle, 1 L
Catalog # MOLE31302764-1L
Marketsource
Supplier:   Molekula
Tween® 80 (Polysorbate)
Catalog # MOLE31302764-1L
Supplier:  Molekula
Marketsource
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Glass bottle, 1 L

CAS Number:  9005-65-6
Price
Inquire For Price
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:13,07664
  • Boiling Pt:∼100 °C (1013 hPa)
  • Melting Pt:–21 °C
  • Density:1.062 g/cm³ (20 °C)
  • Flash Pt:>149 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.