Order Entry
Belgium
ContactUsLinkComponent
Tween® 80 (Polysorbate), BAKER, Laboratory Reagent, J.T.Baker®
Tween® 80 (Polysorbate), BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 7394.0500
Supplier:  Avantor
CAS Number:  
Tween® 80 (Polysorbate), BAKER, Laboratory Reagent, J.T.Baker®
Catalog # 7394.0500
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
Merck Index:13,07664
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:–21 °C
Density:1.062 g/cm³ (20 °C)
Flash Pt:>149 °C
Storage Temperature:Ambient
EINECS:500-019-9
CAS Number:9005-65-6
MDL Number:MFCD00082107