Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Paraformaldehyde

Flammable
Health
Irritant
Marketsource
Supplier:   Serva
Paraformaldehyde
Supplier:  Serva
Marketsource
Certificates
Paraformaldehyde

2 Options Available Below

Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 mmHg)
  • Melting Pt:123 °C
  • Density:1.46 g/cm³ (20 °C)
  • Flash Pt:70 °C
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 20,00 €
    20,00 €
    1 * 100 g
       20,00 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 39,00 €
    39,00 €
    1 * 500 g
       39,00 €

    Specifications:
    Marketsource
    Marketsource