Order Entry
Belgium
Orders LinkContactUsLinkComponent
Paraformaldehyde, powder, TECHNICAL

Paraformaldehyde, powder, TECHNICAL

Flammable
Health
Irritant
Supplier:   VWR Chemicals
Paraformaldehyde, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Paraformaldehyde, powder, TECHNICAL

3 Options Available Below

Product Details & Documents
Synonyms:  Polyoxymethylene
  • CAS Number:30525-89-4
  • Formula:(CH₂O)n
  • MDL Number:MFCD00133991
  • Merck Index:13,07096
  • Boiling Pt:135…136 °C (13 Torr)
  • Melting Pt:120…170 °C
  • Density:0.88 g/cm³ (25 °C)
  • Flash Pt:70 °C (closed cup)
  • Storage Temperature:Ambient
  • UN:2213
  • ADR:4.1,III
Specifications for Product Family

Specification Test Results

  • Identification:
    Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.