Order Entry
Belgium
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # PIER28314
CAS Number:  
undefined
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # PIER28314
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Seq:
    0125
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
Merck Index:13,06793
Boiling Pt:∼200 °C (1013 hPa)
Melting Pt:6 °C
Density:1.06 g/cm³ (20 °C)
Flash Pt:251 °C (closed cup)
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Refrigerator
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505