Polyethylene glycol 6,000 for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25322-68-3
:MFCD00081839
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD01779614
:6000 g/mol
Boiling Pt:∼250 °C (1013 hPa)
Melting Pt:55…62 °C
Density:1.13 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-038-2
Specification Test Results
- Solubility:500 g/l (20 °C)
- Melting Point:55 - 60 °C
- Molar Mass:8000 g/mol
- Vapor Pressure:- 0.01 hPa (20 °C)
- Flash Point:250 °C
- pH:5 - 7 (100 g/l, H2O, 20 °C)
- Ignition Point:420 °C
- Viscosity kinematic:200 - 250 mm2/s (20 °C) (50% solution)
Specification Test Results
Related Information
Frequently Bought Together