Polyethylene glycol 20,000, EMPROVE® ESSENTIAL Ph. Eur. suitable for use as excipient, SAFC®
Catalog # 8.17018.1000
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
- Environmentally Preferable:
Specifications
About this item
Synonyms: PEG 20000, Ethylene glycol polymer 20000, PEG 20,000
:
:20000 g/mol
Melting Pt:58…63 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:500-038-2
CAS Number:25322-68-3
Formula:H(OCH₂CH₂)nOH
MDL Number:MFCD00081839