Order Entry
Belgium
ContactUsLinkComponent
Barium standard solution, concentrate, 1000 mg Ba in 7% hydrochloric acid (from BaCl₂) Titrisol® standard for AAS, Supelco®
Barium standard solution, concentrate, 1000 mg Ba in 7% hydrochloric acid (from BaCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09968.0001
Supplier:  Merck
CAS Number:  
Barium standard solution, concentrate, 1000 mg Ba in 7% hydrochloric acid (from BaCl₂) Titrisol® standard for AAS, Supelco®
Catalog # 1.09968.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg
  • Matrix:
    7% HCl
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Barium
  • Symbol:
    Ba

Specifications

About this item

BaCl₂ in 7% HCl

Certifications: Traceable to SRM from NIST.