Order Entry
Belgium
ContactUsLinkComponent
 
Aluminium standard solution, 10000 mg/L Al in dil. nitric acid (from Al(NO₃)₃), Certipur® standard for ICP, Supelco®
Catalog # 1.70371.0100
Supplier:  Merck
CAS Number:  
Aluminium standard solution, 10000 mg/L Al in dil. nitric acid (from Al(NO₃)₃), Certipur® standard for ICP, Supelco®
Catalog # 1.70371.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Certifications: Traceable to SRM from NIST.