Order Entry
Belgium
ContactUsLinkComponent
Aluminium standard solution, concentrate, 1000 mg Al in water (from AlCl₃) Titrisol® standard for AAS, Supelco®
Aluminium standard solution, concentrate, 1000 mg Al in water (from AlCl₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09967.0001
Supplier:  Merck
CAS Number:  
Aluminium standard solution, concentrate, 1000 mg Al in water (from AlCl₃) Titrisol® standard for AAS, Supelco®
Catalog # 1.09967.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Aluminium
  • Symbol:
    Al

Specifications

About this item

Certifications: Traceable to SRM from NIST.