Order Entry
Belgium
ContactUsLinkComponent
Benzene-[D6] 2000 µg/ml in methanol
Benzene-[D6] 2000 µg/ml in methanol
Supplier:  Restek
Certificates
Benzene-[D6] 2000 µg/ml in methanol
Supplier:  Restek

Specifications

  • CAS No.:
    1076-43-3
  • Formula:
    C₆D₆
  • Description:
    Benzene-[D6]

Specifications

Frequently Bought Together