Glucose oxidase (from Aspergillus niger)
Supplier: Serva
Certificates
About this item
Specifications
- CAS No.:9001-37-0
- Source:Aspergillus niger
- Storage conditions:Freezer
- MDL No.:MFCD00131182
- Enzyme name:Glucose oxidase
Specifications
Frequently Bought Together