Order Entry
Belgium
ContactUsLinkComponent
Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution
Toxic
Health
Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution
Catalog # A3658.1000
Supplier:  PanReac AppliChem
CAS Number:  
Toxic
Health
Acrylamide : Bisacrylamide (19:1) 40% in aqueous solution
Catalog # A3658.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Density:1.03 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3426
ADR:6.1,III
 :GHS06
 :GHS08
 :79-06-1
MDL Number:MFCD00080848