Order Entry
Belgium
ContactUsLinkComponent
Lactic acid ≥90% in aqueous solution
Corrosive
Lactic acid ≥90% in aqueous solution
Catalog # ROTH8994.1
Supplier:  Roth Carl
CAS Number:  
Corrosive
Lactic acid ≥90% in aqueous solution
Catalog # ROTH8994.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Boiling Pt:122 °C (20 hPa)
Density:1.11…1.21 g/cm³ (20 °C)
 :90.08 g/mol
EINECS:200-018-0
 :GHS05
Storage Temperature:Ambient
CAS Number:50-21-5
 :598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520