Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agar, powder, TECHNICAL

Agar, powder, TECHNICAL

Agar, powder, TECHNICAL
Supplier:  VWR Chemicals
Certificates
Agar, powder, TECHNICAL

3 Options Available Below

Product Details & Documents

Not suitable for microbiological purposes

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Specifications for Product Family

Specification Test Results

  • Identification:
    Passes test
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.