Order Entry
Belgium
Orders LinkContactUsLinkComponent
Agar for bacteriology

Agar for bacteriology

Agar for bacteriology
Supplier:  VWR Chemicals
Certificates
Agar for bacteriology

3 Options Available Below

Product Details & Documents

Agar, a solidifying agent used in bacteriology which consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.

  • Agar for the growth of bacteria and fungi
  • Typically used at concentrations of 1 to 2% in solid media preparation
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1

Documents

Specifications for Product Family

Specification Test Results

  • Ash :
    ≤6.5 %
  • Gel Point (1.5%):
    32 – 38 °C
  • Gel Strength (1.5%):
    550 – 950 g/cm²
  • Heavy Metals:
    ≤0.002
  • Loss on Drying:
    ≤12 %
  • Melting Point:
    80 – 90 °C
  • Pass Thru 60 Mesh:
    ≥90 %
  • Turbidity (Nephelos):
    ≤40
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.