TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) 10% in water
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
CAS Number:9002-93-1
:9036-19-5
MDL Number:MFCD00132505
Boiling Pt:∼100 °C (1013 hPa)
Density:∼1.01 g/cm³ (20 °C)
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together