L(+)-Arabinose
Catalog # ROTH5118.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:150.13 g/mol
Boiling Pt:331 °C (1013 hPa)
Melting Pt:158…160 °C
Density:1.585 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:226-214-6
CAS Number:5328-37-0
:87-72-9
Formula:C₅H₁₀O₅
MDL Number:MFCD00067709