Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Mannose

D-(+)-Mannose

Marketsource
Supplier:   NEN LIFE SCIENCE
D-(+)-Mannose
Supplier:  NEN LIFE SCIENCE
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,05772
  • Boiling Pt:376 °C (1013 hPa)
  • Melting Pt:133 °C
  • Density:1.539 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:222-392-4
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 1
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un.

    Specifications:
    Marketsource
    Marketsource
  • 1
    Log in for availability
    Log in for availability
    1 * 1 Un.
    1 * 1 Un.

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.