
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:14,04459
Melting Pt:135…152 °C
Density:1.54 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:200-075-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Meets E.P. Chemical Specifications:—
- Meets U.S.P Requirements:—
- GMP Manufactured Product:—
- Meets B.P. Chemical Specifications:—
- Meets J.P. Chemical Specifications:—
- Preserve in Tight Containers:—
- CAUTION: For Manufacturing, processing or repackaging:—
- Bulk Pharmaceutical Chemical:—
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification:Passes Test
- USP - Water (H₂O):7.5 - 9.0 %
- USP - Readily Carbonizable Substances:Passes Test
- USP - Residue on Ignition:≤0.1 %
- USP - Sulfate (SO₄):≤0.015 %
- USP - Oxalic Acid:≤0.036 %
- USP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Readily Carbonizable Substances:Passes Test
- EP/BP - Oxalic Acid:≤360 ppm
- EP/BP - Sulfate (SO₄):≤150 ppm
- EP/BP - Water (H₂O):7.5 - 9.0 %
- EP/BP - Ash (sulfated):≤0.1 %
- EP/BP - Endotoxin Concentration, IU/mg:≤0.5
- JP - Assay (C₆H₈O₇) (anhydrous basis):99.5 - 100.5 %
- JP - Identification:Passes Test
- JP - Sulfate (SO₄):≤150 ppm
- JP - Oxalic Acid:≤360 ppm
- JP - Clarity and Color of Solution:Passes Test
- JP - Heavy Metals (as Pb):≤10 ppm
- JP - Water (H₂O):7.5 - 9.0 %
- JP - Readily Carbonizable Substances:Passes Test
- JP - Residue on Ignition:≤0.10 %
Specification Test Results
Frequently Bought Together