Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

D-(+)-Galactose

Marketsource
Supplier:   Roth Carl
D-(+)-Galactose
Supplier:  Roth Carl
Marketsource
Certificates
D-(+)-Galactose

2 Options Available Below

Product Details & Documents
Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04356
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:163…169 °C
  • Density:1.5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource