Orders LinkContactUsLinkComponent
D-(+)-Galactose, Millipore®
Glass bottle for solids, 100 g
D-(+)-Galactose, Millipore®
  3455-100
 :  59-23-4
D-(+)-Galactose, Millipore®
D-(+)-Galactose, Millipore®
Glass bottle for solids, 100 g
 :  59-23-4
1 * 100 g
 :  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  •  :59-23-4
  •  :C₆H₁₂O₆
  •  :MFCD00151230
  •  :180.16 g/mol
  •  :13,04356
  •  :527 °C (1013 hPa)
  •  :163…169 °C
  •  :1.5 g/cm³ (20 °C)
  •  :Ambient
  •  :200-416-4
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g