Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Galactose ≥98%

D-(+)-Galactose ≥98%

D-(+)-Galactose ≥98%
Certificates
D-(+)-Galactose ≥98%
D-(+)-Galactose ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 59-23-4 Molecular Formula: C6H12O6 Formula Weight: 180.16 Melting Point: 164-168° Beilstein Reference: 1724619 Merck Reference: 14,4335 Optical Rotation: +80° (c=5 in water, 24h) MDL No.: MFCD00151230

Synonyms:  D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
  • CAS Number:59-23-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00151230
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,4335
  • Boiling Pt:527 °C (1013 hPa)
  • Melting Pt:164…168 °C
  • Density:1.50
  • Storage Temperature:Ambient
  • EINECS:200-416-4
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.