Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Sucrose

D-(+)-Sucrose

D-(+)-Sucrose
Supplier:  PanReac AppliChem
Certificates
D-(+)-Sucrose

2 Options Available Below

Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-334-9

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 65,10 €
    65,10 €
    1 * 1 kg
       65,10 €

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 252,00 €
    252,00 €
    1 * 5 kg
       252,00 €