Order Entry
Belgium
ContactUsLinkComponent
D-(+)-Sucrose octaacetate for synthesis, Sigma-Aldrich®
D-(+)-Sucrose octaacetate for synthesis, Sigma-Aldrich®
Catalog # 8.22144.0250
Supplier:  Merck
CAS Number:  
D-(+)-Sucrose octaacetate for synthesis, Sigma-Aldrich®
Catalog # 8.22144.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    82 - 87 °C
  • Molar Mass:
    678.59 g/mol
  • Boiling Point:
    250 °C (1.3 hPa)
  • Synonyms:
    Octaacetylsucrose
  • Hill Formula:
    C28H38O19

Specification Test Results

About this item

 :
Merck Index:12,09052
Melting Pt:80…83 °C
Density:1.257 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
 :678.6 g/mol
Storage Temperature:Refrigerator
EINECS:204-772-1
CAS Number:126-14-7
Formula:C₂₈H₃₈O₁₉
MDL Number:MFCD00006623