D-(+)-Glucose monohydrate, powder, TECHNICAL
: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
:14431-43-7
:5996-10-1
:C₆H₁₂O₆·H₂O
:MFCD00149450
:198.17 g/mol
:83 °C
:Ambient
:200-075-1
- Environmentally Preferable:
- Water:7.0 - 9.5 %
Frequently Bought Together