Order Entry
Belgium
ContactUsLinkComponent
D-(+)-Glucose monohydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
D-(+)-Glucose monohydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Glucose monohydrate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Chloride (Cl):
    max. 0.005 %
  • Insoluble Matter:
    max. 0.005 %
  • Residue after Ignition:
    max. 0.015 %
  • Starch:
    passes test
  • Sulfate and Sulfite (as SO4):
    max. 0.005 %
  • Trace Impurities (in ppm)Arsenic (As):
    max. 0.2
  • Trace Impurities (in ppm)Iron (Fe):
    max. 5
  • Trace Impurities (in ppm)Heavy Metals (as Pb):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together