Order Entry
Belgium
ContactUsLinkComponent
D-(+)-Glucose monohydrate ≥99.5% for biotechnology
D-(+)-Glucose monohydrate ≥99.5% for biotechnology
Catalog # 0643-1KG
Supplier:  VWR Chemicals
CAS Number:  
undefined
D-(+)-Glucose monohydrate ≥99.5% for biotechnology
Catalog # 0643-1KG
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    HDPE bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • DNase:
    None detected
  • Insolubles:
    ≤0.005%
  • Loss on drying:
    8 – 9%
  • Purity (anhydrous):
    ≥99.5%
  • RNase:
    None detected
  • Specific rotation:
    52.6 – 53.2°

Specification Test Results

About this item

 :
 :198.17 g/mol
Melting Pt:83 °C
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:14431-43-7
 :5996-10-1
Formula:C₆H₁₂O₆·H₂O
MDL Number:MFCD00149450