Order Entry
Belgium
ContactUsLinkComponent
 
D-(+)-Maltose monohydrate ≥95.0% (by HPLC), EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.05911.9029
CAS Number:  
undefined
D-(+)-Maltose monohydrate ≥95.0% (by HPLC), EMPROVE® ESSENTIAL, SAFC®
Catalog # 1.05911.9029
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥ 95.0 %
  • Identity (IR):
    passes test
  • Appearance of solution (clearness):
    passes test
  • Appearance of solution (colour):
    passes test
  • pH-value (5 %; water; 20°C):
    4.5 - 6.0
  • Spec. rotation [α²0/D (10 %; water; calc. on anhydrous substance):
    135.0 - 139.0 °
  • Heavy metals (as Pb):
    ≤ 0.002 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash:
    ≤ 0.1 %
  • Water:
    4.0 - 7.0 %

Specification Test Results

About this item

 :
 :360.32 g/mol
Merck Index:13,05736
Melting Pt:160…165 °C
Storage Temperature:Ambient
EINECS:200-716-5
CAS Number:6363-53-7
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00149343