Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Maltose monohydrate

D-(+)-Maltose monohydrate

D-(+)-Maltose monohydrate
Certificates
D-(+)-Maltose monohydrate

2 Options Available Below

Product Details & Documents
Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • MDL Number:MFCD00149343
  • Molecular Weight:360.32 g/mol
  • Merck Index:13,05736
  • Melting Pt:160…165 °C
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

    Specifications:
    More
    Specifications
    Cat. No.
    BMOL05957-1
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    More
    Specifications
    Cat. No.
    BMOL05957-2