Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Maltose monohydrate

D-(+)-Maltose monohydrate

Glass bottle for solids, 1 kg
Catalog # A3891.1000
Supplier:   PanReac AppliChem
D-(+)-Maltose monohydrate
Catalog # A3891.1000
Supplier:  PanReac AppliChem
CAS Number:  6363-53-7
D-(+)-Maltose monohydrate
D-(+)-Maltose monohydrate

Glass bottle for solids, 1 kg

CAS Number:  6363-53-7
Price
291,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • MDL Number:MFCD00149343
  • Molecular Weight:360.32 g/mol
  • Merck Index:13,05736
  • Melting Pt:160…165 °C
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.