Order Entry
Belgium
Orders LinkContactUsLinkComponent
D-(+)-Maltose monohydrate 95%

D-(+)-Maltose monohydrate 95%

D-(+)-Maltose monohydrate 95%
Certificates
D-(+)-Maltose monohydrate 95%
D-(+)-Maltose monohydrate 95%

2 Options Available Below

Product Details & Documents

Purity: 95% CAS No.: 6363-53-7 Molecular Formula: C12H22O11.H2O Formula Weight: 360.32 (342.30anhy) Melting Point: ca 120° dec. Solubility: Soluble in water. Slightly soluble in alcohol. Insoluble in ether Beilstein Reference: 5784659 Merck Reference: 14,5714 Optical Rotation: +130° (c=2 in water, 24h) MDL No.: MFCD00149343

Synonyms:  Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate
  • CAS Number:6363-53-7
  • Formula:C₁₂H₂₂O₁₁·1H₂O
  • MDL Number:MFCD00149343
  • Molecular Weight:360.32 g/mol
  • Merck Index:14,5714
  • Melting Pt:∼ 120 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:200-716-5
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.