Order Entry
Belgium
ContactUsLinkComponent
Amyloid plaque stain reagent, with EtBr counter stain, Amylo-Glo® RTD™
Amyloid plaque stain reagent, with EtBr counter stain, Amylo-Glo® RTD™
Catalog # BSENTR-400-AG
Supplier:  Biosensis
CAS Number:  
undefined
Amyloid plaque stain reagent, with EtBr counter stain, Amylo-Glo® RTD™
Catalog # BSENTR-400-AG
Supplier:  Biosensis
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Description:
    Amyloid plaque stain reagent with EtBr counter stain

Specifications

About this item

Amylo-Glo RTD reagent is designed to stain amyloid plaques in tissue sections.

  • For Amyloid plaques both intraneuronal and vascular for Amylo-Glo
  • Very bright blue UV excitable stain under physiological conditions
  • Unique excitation/emission profile and mild staining
  • Combination for multiple immunofluorescent labeling studies
  • Storage Up to six months at 4 °C

Amylo-Glo RTD is compatible with fresh, frozen, and formalin-fixed immunohistochemistry or cytochemistry, and particularly good for confocal and multiple labeling because of its high fluorescent intensity and resistance to photo-bleaching.

Moreover because Amylo-Glo fluoresces in the UV channel, double and triple labeling experiments can be performed very easily.

Ethidium bromide is used as a counter stain to visualise cell nuclei and cell bodies of cells while under UV illumination allowing the assessment of amyloid plaques and cell/tissue positioning as well in one step.

Caution: Protect from light. No preservatives. Use sterile technique when handling and proper laboratory procedures.