Petroleum spirit, 40…60 °C, CHROMASOLV™ for pesticide residue analysis, low boiling point hydrogen treated naphtha, Riedel-de Haën™
:64742-49-0
:64742-82-1
:MFCD00081849
:40…60 °C (1.013 hPa)
:0.65 g/cm³ (20 °C)
:–30 °C
:Ambient
:309-852-0
:3295
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.01 %
- Suitability for residue analysis:Max. 5 ng/l
Frequently Bought Together