Burettes, straight stopcock, PTFE spindle, Schellbach, BLAUBRAND®
: Brand
Borosilicate glass 3.3, class AS.
- Calibrated to deliver 'TD, Ex', DE-M marking
- Schellbach stripe, ring marks at major graduations
- Marks and inscriptions in high contrast blue enamel represent the optimum combination of resistance and readability
Length: 800 mm
With individual certificate
: DIN EN ISO 385
Frequently Bought Together