Order Entry
Belgium
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0810.1000
Supplier:  Avantor
CAS Number:  
Molecular sieve A4 (0.4 nm, 4 Å), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 0810.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Loss on Drying at 300°C:
    max. 5 %

Specification Test Results

About this item

 :
Density:1.1…2.2 g/cm³ (20 °C)
CAS Number:70955-01-0
 :1318-02-1
MDL Number:MFCD03457537