Order Entry
Belgium
ContactUsLinkComponent
Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh
Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh
Certificates
undefined
Molecular sieve A4 (0.4 nm, 4 Å) 10 --> 18 mesh

Specification Test Results

  • Appearance:
    Beige Granules
  • Bulk density:
    680 to 780 g/l
  • Residual water:
    ≤1.5 % (50°C, 2 h)
  • Water adsorption:
    ≥20 % (50% rH, 20°C, 24 h)

Specification Test Results

Related Information

Frequently Bought Together