Order Entry
Belgium
ContactUsLinkComponent
Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 3.2 mm, Fluka™
Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 3.2 mm, Fluka™
Catalog # 334278-1KG
CAS Number:  
Molecular sieve 3A (0.3 nm, 3 Å), pellets, Ø 3.2 mm, Fluka™
Catalog # 334278-1KG
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Particle Size:
    3.2 mm
  • Form:
    pellets
  • Granulation:
    complying
  • Pore Size (Ång Ström):
    3

Specification Test Results

About this item

 :
Melting Pt:400 °C
Density:2 g/cm³
Storage Temperature:Ambient
CAS Number:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00147627