Molecular sieve 3A (0.3 nm, 3 Å), beads 8-12 mesh, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00147627
Melting Pt:400 °C
Density:2 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Form:beads
- Grain size 8 - 12 mesh:Min. 95 %
- Pore Size (Ång Ström):3
Specification Test Results
Frequently Bought Together