
Molecular sieve 3A (0.3 nm, 3 Å), powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:308080-99-1
:1318-02-1
Formula:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number:MFCD00131613
:GHS07
Storage Temperature:Ambient
Frequently Bought Together