Molecular sieve 3A (0.3 nm, 3 Å) 4 --> 8 mesh
:308080-99-1
:1318-02-1
:KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
:MFCD00131613
- Appearance:Beige to tan Beads
- Particle size:4 to 8 mesh
- Bulk density:≥600 g/l
- Residual water:≤1.5 %
- Water adsorption:≥20 % (60% rH)
Frequently Bought Together