


Xylene (mixture of isomers) for histology
Catalog # 534056-4L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Aluminium bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Vapor Pressure:18 mmHg ( 37.7 °C)
- IR spectrum:complying
- C₈ isomers including Ethylbenzene (GC):Min. 96 %
- Total isomers (GC):Min. 73 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Ethyl benzene (GC):Max. 25.0 %
- APHA:Max. 10
Specification Test Results
About this item
:
:106.17 g/mol
Merck Index:13,10136
Boiling Pt:136 - 140 °C
Boiling Pt:136…140 °C
Melting Pt:-34 °C
Melting Pt:–34 °C
Density:0.87 g/cm³
Flash Pt:25 °C
EINECS:215-535-7
UN:1307
ADR:3,III
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264