Order Entry
Belgium
Orders LinkContactUsLinkComponent
Iron ≥98%, powder, Technical Grade

Iron ≥98%, powder, Technical Grade

Plastic bottle for solids, 1 kg
Catalog # 24086.292
Flammable
Supplier:   VWR Chemicals
Iron ≥98%, powder, Technical Grade
Catalog # 24086.292
Supplier:  VWR Chemicals
CAS Number:  7439-89-6
Iron ≥98%, powder, Technical Grade
Iron ≥98%, powder, Technical Grade

Plastic bottle for solids, 1 kg

CAS Number:  7439-89-6
Price
78,10 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:7439-89-6
  • Formula:Fe
  • MDL Number:MFCD00010999
  • Molecular Weight:55.85 g/mol
  • Boiling Pt:3000 °C (1013 hPa)
  • Melting Pt:1536 °C
  • Density:7.86 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-096-4
  • UN:3089
  • ADR:4.1,III
Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.